missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benedict's Reagent, Pure, Quantitative, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Specifications
| MDL Number | 146068 |
| Packaging | Amber glass bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- Benedict's reagent
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
- MIXTURE LIST-Contains :
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction