Learn More
Benaprizine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Benaprizine Hydrochloride |
| CAS | 3202-55-9 |
| Analyte or Component Names | Benaprizine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzeneacetic acid, α-hydroxy-α-phenyl-, 2-(ethylpropylamino)ethyl ester, hydrochloride (9CI), Benzilic acid, 2-(ethylpropylamino)ethyl ester hydrochloride (7CI,8CI), Ethanol, 2-(ethylpropylamino)-, benzilate (ester), hydrochloride (8CI), 2-(Ethyl-n-propylamino)ethyl α,α-diphenylglycolate hydrochloride, 2-(Ethylpropylamino)ethyl benzilate hydrochloride, AP 1288, BRL 1288, Benapryzine hydrochloride, Benzeneacetic acid, α-hydroxy-α-phenyl-, 2-(ethylpropylamino)ethyl ester, hydrochloride (1:1) |
| InChI Formula | InChI=1S/C21H27NO3.ClH/c1-3-15-22(4-2)16-17-25-20(23)21(24,18-11-7-5-8-12-18)19-13-9-6-10-14-19;/h5-14,24H,3-4,15-17H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CCCN(CC)CCOC(=O)C(O)(c1ccccc1)c2ccccc2 |
| Show More |