Learn More
BB-K 6 N-(Benzyloxy) Carbamate, TRC
Product Code. 30340996
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340996 100 mg 100mg
30339641 10 mg 10mg
30333210 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30340996 Supplier Toronto Research Chemicals Supplier No. TRCB119105100MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material BB-K 6 N-(Benzyloxy) Carbamate
Analyte or Component Names BB-K 6 N-(Benzyloxy) Carbamate
Shipping Condition Room Temperature
Molecular Formula C30 H49 N5 O15
InChI Formula InChI=1S/C30H49N5O15/c31-13-8-14(32)26(24(43)25(13)49-28-21(40)18(33)19(38)17(10-36)48-28)50-29-23(42)22(41)20(39)16(47-29)9-35-27(44)15(37)6-7-34-30(45)46-11-12-4-2-1-3-5-12/h1-5,13-26,28-29,36-43H,6-11,31-33H2,(H,34,45)(H,35,44)/t13-,14+,15+,16-,17-,18+,19-,20-,21-,22+,23-,24-,25+,26-,28-,29-/m1/s1
Solution Type Neat
SMILES N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CNC(=O)[C@@H](O)CCNC(=O)OCc3ccccc3)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]4O[C@H](CO)[C@@H](O)[C@H](N)[C@H]4O
IUPAC Name benzyl N-[(3S)-4-[[(2R,3S,4S,5R,6R)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-3,4,5-trihydroxyoxan-2-yl]methylamino]-3-hydroxy-4-oxobutyl]carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 719.735
Quantity 100 mg
Formula Weight 719.323