Learn More
Baquiloprim, TRC
Product Code. 17924495
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17924495 25 mg 25mg
17962655 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17924495 Supplier Toronto Research Chemicals Supplier No. TRCB11800025MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102280-35-3
Molecular Formula C17 H20 N6
Molecular Weight (g/mol) 308.38
Synonym 5-[[8-(Dimethylamino)-7-methyl-5-quinolinyl]methyl]-2,4-pyrimidinediamine, 138OU, Baquiloprim, 2,4-Pyrimidinediamine, 5-[[8-(dimethylamino)-7-methyl-5-quinolinyl]methyl]-
IUPAC Name 5-[[8-(dimethylamino)-7-methylquinolin-5-yl]methyl]pyrimidine-2,4-diamine
SMILES CN(C)c1c(C)cc(Cc2cnc(N)nc2N)c3cccnc13
Specifications
Analyte or Component Names Baquiloprim
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H20N6/c1-10-7-11(8-12-9-21-17(19)22-16(12)18)13-5-4-6-20-14(13)15(10)23(2)3/h4-7,9H,8H2,1-3H3,(H4,18,19,21,22)
Solution Type Neat
Industry Type Forensics
Quantity 25 mg
Formula Weight 308.1749
Percent Purity >95%
Chemical Name or Material Baquiloprim
Grade HPLC