Learn More
AZD2858, MedChemExpress
Product Code. 16432877
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16432877
Brand: Medchem Express HY15761
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress AZD2858 is a potent, orally active GSK-3 inhibitor, with IC50s of 0.9 and 5 nM for GSK-3α and GSK-3β, respectively, used in the research of fracture healing.
AZD2858 is a potent, orally active GSK-3 inhibitor, with IC50s of 0.9 and 5 nM for GSK-3α and GSK-3β, respectively, used in the research of fracture healing.
Specifications
Chemical Name or Material AZD2858
CAS 486424-20-8
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C21H23N7O3S
Quantity 50 mg
Solubility Information DMSO : 12.5 mg/mL (27.56 mM; Need ultrasonic)
SMILES O=C(C1=NC(C2=CC=C(S(=O)(N3CCN(C)CC3)=O)C=C2)=CN=C1N)NC4=CC=CN=C4
Molecular Weight (g/mol) 453.52
Formula Weight 453.52
Percent Purity 98.42%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation