Learn More
Axitinib, Mikromol
Product Code. 30461449
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461449 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461449 Supplier Mikromol Supplier No. MM3922.000250
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Axitinib, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Axitinib
CAS 319460-85-0
Analyte or Component Names Axitinib
Shipping Condition Room Temperature
Molecular Formula C22 H18 N4 O S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C22H18N4OS/c1-23-22(27)18-7-2-3-8-21(18)28-16-10-11-17-19(25-26-20(17)14-16)12-9-15-6-4-5-13-24-15/h2-14H,1H3,(H,23,27)(H,25,26)/b12-9+
Solution Type Neat
SMILES CNC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)n[nH]c3c2
IUPAC Name N-methyl-2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]benzamide
Molecular Weight (g/mol) 386.47
Quantity 250 mg
Formula Weight 386.1201 g/mol