missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRIP Antibody (SR2262), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 449.00 €
Specifications
| Antigen | ATRIP |
|---|---|
| Clone | SR2262 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650165
|
Novus Biologicals
NBP3-22394-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18605534
|
Novus Biologicals
NBP3-22394-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATRIP Monoclonal antibody specifically detects ATRIP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| ATRIP | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
| A synthesized peptide derived from human ATRIP (Uniprot #: Q8WXE1) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2262 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 84126 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title