missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRIP Antibody (SR2262), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22394-50ul
This item is not returnable.
View return policy
Description
ATRIP Monoclonal antibody specifically detects ATRIP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| ATRIP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR2262 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
| A synthesized peptide derived from human ATRIP (Uniprot #: Q8WXE1) | |
| 50 μg | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| 84126 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction