Learn More
Artemisitene, TRC
Product Code. 30005820
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005820 25 mg 25mg
30010053 50 mg 50mg
30006223 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005820 Supplier Toronto Research Chemicals Supplier No. TRCA77755025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101020-89-7
Molecular Formula C15 H20 O5
Molecular Weight (g/mol) 280.32
Synonym (3R,5aS,6R,8aS,12S,12aR)-Octahydro-3,6-dimethyl-9-methylene-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10(3H)-one, (+)-Artemisitene, Dehydroqinghaosu;
IUPAC Name (1R,4S,5R,8S,12S,13R)-1,5-dimethyl-9-methylidene-11,14,15,16-tetraoxatetracyclo[10.3.1.04,13.08,13]hexadecan-10-one
SMILES C[C@@H]1CC[C@H]2C(=C)C(=O)O[C@@H]3O[C@@]4(C)CC[C@@H]1[C@@]23OO4
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C15H20O5/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-20-15/h8,10-11,13H,2,4-7H2,1,3H3/t8-,10+,11+,13-,14-,15-/m1/s1
Formula Weight 280.1311
Percent Purity >95%
Chemical Name or Material Artemisitene
Grade HPLC