missing translation for 'onlineSavingsMsg'
Learn More

ARID2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

226.00 € - 463.00 €

Specifications

Antigen ARID2
Dilution Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18686150
View Documents
Novus Biologicals
NBP2-92943-0.02ml
0.02 mL
226.00 €
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18605021
View Documents
Novus Biologicals
NBP2-92943-0.1ml
0.1 mL
463.00 €
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

ARID2
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Chromatin Modifiers
PBS with 50% glycerol, pH7.3.
196528
IgG
Affinity purified
Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000
Polyclonal
Purified
RUO
Human
ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200
Recombinant fusion protein containing a sequence corresponding to amino acids 1586-1835 of human ARID2 (NP_689854.2). QNVVPQNTPMPPSPAVQVQGQPNSSQPSPFSGSSQPGDPMRKPGQNFMCLWQSCKKWFQTPSQVFYHAATEHGGKDVYPGQCLWEGCEPFQRQRFSFITHLQDKHCSKDALLAGLKQDEPGQAGSQKSSTKQPTVGGTSSTPRAQKAIVNHPSAALMALRRGSRNLVFRDFTDEKEGPITKHIRLTAALILKNIGKYSECGRRLLKRHENNLSVLAISNMEASSTLAKCLYELNFTVQSKEQEKDSEMLQ
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.