Learn More
ARID2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92943-0.1ml
This item is not returnable.
View return policy
Description
ARID2 Polyclonal antibody specifically detects ARID2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ARID2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000 | |
| ARID domain-containing protein 2, AT rich interactive domain 2 (ARID, RFX-like), BAF200KIAA1557AT-rich interactive domain-containing protein 2, BRG1-associated factor 200, DKFZp686G052, DKFZp779P0222, FLJ30619, p200, Zinc finger protein with activation potential, zipzap, zipzap/p200 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1586-1835 of human ARID2 (NP_689854.2). QNVVPQNTPMPPSPAVQVQGQPNSSQPSPFSGSSQPGDPMRKPGQNFMCLWQSCKKWFQTPSQVFYHAATEHGGKDVYPGQCLWEGCEPFQRQRFSFITHLQDKHCSKDALLAGLKQDEPGQAGSQKSSTKQPTVGGTSSTPRAQKAIVNHPSAALMALRRGSRNLVFRDFTDEKEGPITKHIRLTAALILKNIGKYSECGRRLLKRHENNLSVLAISNMEASSTLAKCLYELNFTVQSKEQEKDSEMLQ | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 196528 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |