Learn More
Antazoline EP Imputiry A (as Dihydrochloride), Mikromol™
Description
Antazoline EP Imputiry A (as Dihydrochloride)
Specifications
Specifications
| Chemical Name or Material | N-(2-Aminoethyl)-2-(benzylphenylamino)acetamide Dihydrochloride |
| CAS | 2724689-85-2 |
| Analyte or Component Names | N-(2-Aminoethyl)-2-(benzylphenylamino)acetamide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H21 N3 O . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Antazoline Hydrochloride Imp. A (EP), Antazoline Imp. A (EP), Antazoline USP Related Compound A, Antazoline USP RC A, N-(2-Aminoethyl)-2-(benzylphenylamino)acetamide Dihydrochloride, Antazoline Hydrochloride Imp. A (EP) as Dihydrochloride, Antazoline Related Compound A as Dihydrochloride, Antazoline Hydrochloride Impurity A as Dihydrochloride, Antazoline Impurity A as Dihydrochloride |
| InChI Formula | InChI=1S/C17H21N3O.2ClH/c18-11-12-19-17(21)14-20(16-9-5-2-6-10-16)13-15-7-3-1-4-8-15;;/h1-10H,11-14,18H2,(H,19,21);2*1H |
| SMILES | Cl.Cl.NCCNC(=O)CN(Cc1ccccc1)c2ccccc2 |
| IUPAC Name | N-(2-aminoethyl)-2-(N-benzylanilino)acetamide;dihydrochloride |
| Show More |