missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | ANO7 |
|---|---|
| Concentration | 0.22 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16879004
|
Proteintech
19486-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16869004
|
Proteintech
19486-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity. May play a role in cell-cell interactions. .Specifications
| ANO7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q6IWH7 | |
| 50636 | |
| Peptide | |
| Primary | |
| -20°C | |
| ANO7 |
| 0.22 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ANO7, anoctamin 7, D TMPP, DTMPP, IPCA 5, IPCA5, New gene expressed in prostate, NGEP, PCANAP5, PCANAP5L, TMEM16G, Transmembrane protein 16G | |
| ANO7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title