missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANO7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19486-1-AP-20UL
This item is not returnable.
View return policy
Description
Has calcium-dependent phospholipid scramblase activity; scrambles phosphatidylserine, phosphatidylcholine and galactosylceramide. Does not exhibit calcium-activated chloride channel (CaCC) activity. May play a role in cell-cell interactions. .Specifications
| ANO7 | |
| Polyclonal | |
| Unconjugated | |
| ANO7 | |
| ANO7, anoctamin 7, D TMPP, DTMPP, IPCA 5, IPCA5, New gene expressed in prostate, NGEP, PCANAP5, PCANAP5L, TMEM16G, Transmembrane protein 16G | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50636 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6IWH7 | |
| ANO7 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction