Learn More
Androsta-1,4-diene-3,17-dione (Androstadienedione), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Androsta-1,4-diene-3,17-dione (Androstadienedione) |
| CAS | 897-06-3 |
| Analyte or Component Names | Androsta-1,4-diene-3,17-dione (Androstadienedione) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Androsta-1,4-diene-3,17-dione, Testosterone Imp. G (EP), Androstadienedione, (+)-Androsta-1,4-diene-3,17-dione, 1-Dehydroandrostenedione, Androsta-1,4-dien-3,17-dione, Androstane-1,4-diene-3,17-dione, Boldione, NSC 49080, Exemestane USP Related Compound C CIII, Exemestane USP RC C CIII, Delta1,4-Androstadiene-3,17-dione, Exemestane Related Compound C, Exemestane Impurity D, Testosterone Impurity G |
| InChI Formula | InChI=1S/C19H24O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h7,9,11,14-16H,3-6,8,10H2,1-2H3/t14-,15-,16-,18-,19-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CCC2=O |
| Show More |