Learn More
Thermo Scientific™ AnaeroBox™ 3.5L Box with Lid and Seal
Successfully culture anaerobic and microaerophilic organisms by achieving the right atmospheric conditions using this combination of Jar, Lid and Seal.
208.00 €
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17391830
View Documents
Thermo Scientific™
AB0035A
3.5 L AnaeroBox
208.00 €
Pack of 1
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Successfully culture anaerobic and microaerophilic organisms by achieving the right atmospheric conditions using Thermo Scientific™ Oxoid™ AnaeroBox™ 3.5L Box with Lid and Seal.
The Box is designed to be used with Oxoid™ AnaeroGen™ 3.5L Sachet (AN0035A) and Oxoid™ CampyGen™ 3.5L Sachet (CN0035A).
Rapidly achieve the required gaseous conditions with easy-to-use Oxoid™ AnaeroBox™ 3.5L Box with Lid and Seal.
The AnaeroBox is designed to be used with Oxoid™ AnaeroGen™ 3.5L Sachet (AN0035A) and Oxoid™ CampyGen™ 3.5L Sachet (CN0035A).
The AnaeroGen 3.5L Sachet (AN0035A) produces an anaerobic atmosphere rapidly without the use of water or catalyst. No hydrogen is produced. The oxygen is absorbed rapidly without the build-up of pressure. The final atmosphere contains less than 1% oxygen supplemented with carbon dioxide - ideal for optimum growth of anaerobes and increased colony size.
The CampyGen 3.5L Sachet (CN0035A) produces a microaerophilic atmosphere rapidly without the use of water or catalyst. No hydrogen is produced. The oxygen is absorbed rapidly without the build-up of pressure. The final atmosphere contains an ideal microaerobic atmosphere of 8-9% oxygen and 7-8% carbon dioxide.
Oxoid™ AnaeroGen™ 3.5L Sachet (AN0035A) and Oxoid™ CampyGen™ 3.5L Sachet (CN0035A) are not included.
Videos
SDS
Documents
Documents
Product Certifications