Learn More
Ampicillin EP Impurity K, Mikromol™
Description
Ampicillin EP Impurity K
Specifications
Specifications
| Chemical Name or Material | (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic Acid |
| CAS | 40610-41-1 |
| Analyte or Component Names | (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H17 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic Acid, Ampicillin Sodium Imp. K (EP), Ampicillin Imp. K (EP), Ampicillin Sodium Impurity K, Ampicillin Impurity K |
| InChI Formula | InChI=1S/C13H17NO3/c1-13(2,3)12(17)14-10(11(15)16)9-7-5-4-6-8-9/h4-8,10H,1-3H3,(H,14,17)(H,15,16)/t10-/m1/s1 |
| SMILES | CC(C)(C)C(=O)N[C@@H](C(=O)O)c1ccccc1 |
| IUPAC Name | (2R)-2-(2,2-dimethylpropanoylamino)-2-phenylacetic acid |
| Show More |