Description
- High ionic strength
- Aqueous matrix
- Used as an additive when using an ion selective electrode (ISE) for the measurement of ammonia.
- Suppresses interfering ions, eliminates matrix, hysteresis or liquid junction potentials.
- Eliminates or reduces error due to temperature co-efficient of variation between sample and calibration standard.
- Certified and has guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Packaging | HDPE Bottle GL45 |
| Physical Form | Liquid |
| Concentration | Ammonium 4 M LiCl |
| Quantity | 500 mL |