Learn More
Ambroxol EP Impurity D (as Hydrochloride), Mikromol™
Description
Ambroxol EP Impurity D (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride |
| CAS | 1384955-66-1 |
| Analyte or Component Names | cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 Br2 N2 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ambroxol Hydrochloride Imp. D (EP), Ambroxol Imp. D (EP), cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride, Ambroxol Hydrochloride Imp. D (EP) as Hydrochloride, Ambroxol Hydrochloride Impurity D as Hydrochloride, Ambroxol Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11+; |
| SMILES | Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@H](O)CC2 |
| IUPAC Name | 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol;hydrochloride |
| Show More |