Learn More
Ambroxol EP Impurity D (as Hydrochloride), Mikromol™
Product Code. 17953196
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953196 100 mg 100mg
17972376 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17953196 Supplier Mikromol Supplier No. MM0005.05
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ambroxol EP Impurity D (as Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride
CAS 1384955-66-1
Analyte or Component Names cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H18 Br2 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym Ambroxol Hydrochloride Imp. D (EP), Ambroxol Imp. D (EP), cis-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol Hydrochloride, Ambroxol Hydrochloride Imp. D (EP) as Hydrochloride, Ambroxol Hydrochloride Impurity D as Hydrochloride, Ambroxol Impurity D as Hydrochloride
InChI Formula InChI=1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11+;
SMILES Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@H](O)CC2
IUPAC Name 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol;hydrochloride
Molecular Weight (g/mol) 414.56
Quantity 100 mg
Formula Weight 411.9553 g/mol