missing translation for 'onlineSavingsMsg'
Learn More

Ambenonium dichloride, Tocris Bioscience™

Product Code. 18782644
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782644 10 mg 10mg
18712554 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782644 Supplier Tocris Bioscience Supplier No. 0388/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cholinesterase inhibitor

Extremely potent, selective and rapidly reversible inhibitor of acetylcholinesterase (AChE) (IC50 values are 0.000698 and 8.20 μM at AChE and BChE respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52022-31-8
Molecular Formula C28H42Cl4N4O2
Molecular Weight (g/mol) 608.47
InChI Key DXUUXWKFVDVHIK-UHFFFAOYSA-N
Synonym ambenonium chloride, ambenonium dichloride, ambenonii chloridum, ambestigmin chloride, mytelase, misuran, mysuran, oksazil, oxamizil, oxazylum
PubChem CID 8288
ChEBI CHEBI:2628
IUPAC Name (2-chlorophenyl)methyl-[2-[[2-[2-[(2-chlorophenyl)methyl-diethylazaniumyl]ethylamino]-2-oxoacetyl]amino]ethyl]-diethylazanium;dichloride
SMILES CC[N+](CC)(CCNC(=O)C(=O)NCC[N+](CC)(CC)CC1=CC=CC=C1Cl)CC2=CC=CC=C2Cl.[Cl-].[Cl-]

Specifications

Quantity 10 mg
Formula Weight 608.48
Percent Purity >99%
Chemical Name or Material Ambenonium dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.