CAS RN 52022-31-8
CAS RN 52022-31-8
IUPAC Name:
[(2-chlorophenyl)methyl](2-{[(2-{[(2-chlorophenyl)methyl]diethylazaniumyl}ethyl)carbamoyl]formamido}ethyl)diethylazanium dichloride
Synonyms:
ambenonium chloride cloruro de ambenonio chlorure d'ambenonium ambenonii chloridum ambenonium dichloride 安贝氯铵 ambestigmin chloride
Molecular Weight (g/mol):
608.47
Molecular Formula:
C28H42Cl4N4O2
InChi Key:
DXUUXWKFVDVHIK-UHFFFAOYSA-N
SMILES:
[Cl-].[Cl-].CC[N+](CC)(CCNC(=O)C(=O)NCC[N+](CC)(CC)CC1=CC=CC=C1Cl)CC1=CC=CC=C1Cl
1
–
1
of
1
results
1
Ambenonium dichloride, Tocris Bioscience™
CAS: 52022-31-8 Molecular Formula: C28H42Cl4N4O2 Molecular Weight (g/mol): 608.47 InChI Key: DXUUXWKFVDVHIK-UHFFFAOYSA-N Synonym: ambenonium chloride,ambenonium dichloride,ambenonii chloridum,ambestigmin chloride,mytelase,misuran,mysuran,oksazil,oxamizil,oxazylum PubChem CID: 8288 ChEBI: CHEBI:2628 IUPAC Name: (2-chlorophenyl)methyl-[2-[[2-[2-[(2-chlorophenyl)methyl-diethylazaniumyl]ethylamino]-2-oxoacetyl]amino]ethyl]-diethylazanium;dichloride SMILES: CC[N+](CC)(CCNC(=O)C(=O)NCC[N+](CC)(CC)CC1=CC=CC=C1Cl)CC2=CC=CC=C2Cl.[Cl-].[Cl-]