Learn More
Alprostadil, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Alprostadil
Specifications
Specifications
| Chemical Name or Material | Alprostadil |
| CAS | 745-65-3 |
| Analyte or Component Names | Alprostadil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H34 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Alprostadil, 7-[(1R,2R,3R)-3-Hydroxy-2-[(1E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]heptanoic acid |
| InChI Formula | InChI=1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1 |
| SMILES | CCCCC[C@H](O)\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)O |
| IUPAC Name | 7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]heptanoic acid |
| Show More |