Learn More
Alpha-Phenyl-Alpha-(2-propyn-1-yloxy)benzeneacetic Acid, TRC
Product Code. 30413266
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413266 1 g
30423686 250 mg
30413581 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30413266 Supplier Toronto Research Chemicals Supplier No. TRCP3364051G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2766-01-0
Molecular Formula C17H14O3
Molecular Weight (g/mol) 266.29
Synonym Diphenyl(2-propynyloxy)acetic Acid, Propinoxiphenyl Mandelic Acid
IUPAC Name 2,2-diphenyl-2-prop-2-ynoxyacetic acid
SMILES OC(=O)C(OCC#C)(c1ccccc1)c2ccccc2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C17H14O3/c1-2-13-20-17(16(18)19,14-9-5-3-6-10-14)15-11-7-4-8-12-15/h1,3-12H,13H2,(H,18,19)
Formula Weight 266.0943
Chemical Name or Material alpha-Phenyl-alpha-(2-propyn-1-yloxy)benzeneacetic Acid