CAS RN 2766-01-0
CAS RN 2766-01-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Phenyl-Alpha-(2-propyn-1-yloxy)benzeneacetic Acid, TRC
CAS: 2766-01-0 Molecular Formula: C17H14O3 Molecular Weight (g/mol): 266.29 Synonym: Diphenyl(2-propynyloxy)acetic Acid,Propinoxiphenyl Mandelic Acid IUPAC Name: 2,2-diphenyl-2-prop-2-ynoxyacetic acid SMILES: OC(=O)C(OCC#C)(c1ccccc1)c2ccccc2