Learn More
alpha,alpha,alpha-Tribromoquinaldine, 97%, Thermo Scientific™
Product Code. 11410728
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11410728
Brand: Thermo Scientific Alfa Aesar B25218.09
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 613-53-6
Molecular Formula C10H6Br3N
Molecular Weight (g/mol) 379.88
MDL Number MFCD00006755
InChI Key UDYYQHILRSDDMP-UHFFFAOYSA-N
PubChem CID 69178
IUPAC Name 2-(tribromomethyl)quinoline
SMILES BrC(Br)(Br)C1=CC=C2C=CC=CC2=N1
Specifications
Melting Point 128°C to 131°C
Quantity 10 g
Beilstein 147638
Synonym 2-tribromomethyl quinoline, quinoline, 2-tribromomethyl, alpha,alpha,alpha-tribromoquinaldine, 2-alpha,alpha,alpha-tribromomethyl quinoline, .alpha.,.alpha.,.alpha.-tribromoquinaldine, acmc-209mrr, 2-tribrommethyl-chinolin, 2-tribromo-methyl-quinoline, quinoline,2-tribromomethyl, udyyqhilrsddmp-uhfffaoysa
Formula Weight 379.89
Percent Purity 97%
Chemical Name or Material alpha,alpha,alpha-Tribromoquinaldine