accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrBrBrN

CAS RN 613-53-6

IUPAC Name: 2-(tribromomethyl)quinoline
Synonyms: quinoline, 2-(tribromomethyl)-
Molecular Weight (g/mol): 379.88
Molecular Formula: C10H6Br3N
InChi Key: UDYYQHILRSDDMP-UHFFFAOYSA-N
SMILES: BrC(Br)(Br)C1=CC=C2C=CC=CC2=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results