missing translation for 'onlineSavingsMsg'
Learn More

alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane, 99%

Product Code. 15497947
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15497947

Brand: Thermo Scientific Alfa Aesar H51066.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 167482-98-6 | C19H22N2 | 278.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167482-98-6
Molecular Formula C19H22N2
Molecular Weight (g/mol) 278.399
MDL Number MFCD03454220
InChI Key SLPZWMKFAWAFSE-UHFFFAOYSA-N
Synonym 5-4-tert-butylphenyl dipyrromethane, alpha-4-tert-butylphenyl di 2-pyrrolyl methane, e'-4-tert-butylphenyl di 2-pyrrolyl methane, 2-4-tert-butylphenyl 1h-pyrrol-2-yl methyl-1h-pyrrole, a-4-tert-butylphenyl di 2-pyrrolyl methane, 2,2'-4-tert-butylphenyl methylenebispyrrole, 2,2'-4-tert-butylbenzylidene bis 1h-pyrrole, 2,2'-4-tert-butylphenyl methylene di 1h-pyrrole, 2-4-tert-butylphenyl-1h-pyrrol-2-yl methyl-1h-pyrrole, 1h-pyrrole, 2,2'-4-1,1-dimethylethyl phenyl methylene bis
PubChem CID 4245355
IUPAC Name 2-[(4-tert-butylphenyl)-(1H-pyrrol-2-yl)methyl]-1H-pyrrole
SMILES CC(C)(C)C1=CC=C(C=C1)C(C2=CC=CN2)C3=CC=CN3

Specifications

Melting Point 158°C to 162°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 278.39
Percent Purity 99%
Chemical Name or Material alpha-(4-tert-Butylphenyl)di(2-pyrrolyl)methane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.