accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 167482-98-6

H3CH3CCH3HNNH

CAS RN 167482-98-6

IUPAC Name: 2-[(4-tert-butylphenyl)(1H-pyrrol-2-yl)methyl]-1H-pyrrole
Synonyms: 2-[(4-tert-butylphenyl)(1H-pyrrol-2-yl)methyl]-1H-pyrrole
Molecular Weight (g/mol): 278.4
Molecular Formula: C19H22N2
InChi Key: SLPZWMKFAWAFSE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(C1=CC=CN1)C1=CC=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results