missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grace™ Alltech™ Vydac™ 214TP C4 Column
A polymerically bonded endcapped n-butyl reversed-phase based on 300Å TP silica
Brand: Grace™ 214TP5115
Additional Details : Weight : 0.10000kg
Description
- Polymeric bonded phases have exceptionally long column lifetime and negligible phase leaching
- Extensive applications library based on over two decade's experience
Specifications
1 mm | |
150 mm | |
5 μm | |
60 to 110 m2/g | |
214 TP | |
3% | |
Yes |
Glycoproteins, Hemoglobin Variants, Histones, Human Growth Hormone, Insulin Variants, Membrane Proteins, Analysis and Purification of Proteins and Peptides | |
Spherical | |
300 Å | |
L26 | |
Silica | |
C4 | |
Column |