Learn More
Alfuzosin EP Impurity D (as Dihydrochloride), Mikromol™
Description
Alfuzosin EP Impurity D (as Dihydrochloride)
Specifications
Specifications
| Chemical Name or Material | N-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-N-methylpropane-1,3-diamine Dihydrochloride |
| CAS | 2732347-83-8 |
| Analyte or Component Names | N-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-N-methylpropane-1,3-diamine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H21 N5 O2 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Alfuzosin Hydrochloride Imp. D (EP), Alfuzosin Imp. D (EP), N-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-N-methylpropane-1,3-diamine Dihydrochloride, Alfuzosin Hydrochloride Imp. D (EP) as Dihydrochloride, Alfuzosin Hydrochloride Impurity D as Dihydrochloride, Alfuzosin Impurity D as Dihydrochloride |
| InChI Formula | InChI=1S/C14H21N3O.2ClH/c1-17-10-5-8-13(9-11-17)15-16-14(18)12-6-3-2-4-7-12;;/h2-4,6-7,13,15H,5,8-11H2,1H3,(H,16,18);2*1H |
| SMILES | Cl.Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCN |
| IUPAC Name | 2-N-(3-aminopropyl)-6,7-dimethoxy-2-N-methylquinazoline-2,4-diamine;dihydrochloride |
| Show More |