Learn More
Albendazole Sulfoxide, TRC
Product Code. 17993985
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993985 50 mg 50mg
17941675 100 mg 100mg
17912975 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17993985 Supplier Toronto Research Chemicals Supplier No. TRCA51162050MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54029-12-8
Molecular Formula C12 H15 N3 O3 S
Molecular Weight (g/mol) 281.33
Synonym Carbamic acid, [5-(propylsulfinyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), (±)-Albendazole sulfoxide, Albendazole oxide, Albendazole sulfoxide, Methyl [5-(propylsulfinyl)-1H-benzimidazol-2-yl]carbamate, RS 8852, Ricobendazole, Rycobendazole
IUPAC Name methyl N-(5-propylsulfinyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCS(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1
Specifications
Analyte or Component Names Albendazole sulfoxide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C12H15N3O3S/c1-3-6-19(17)8-4-5-9-10(7-8)14-11(13-9)15-12(16)18-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 281.0834
Percent Purity >95%
Chemical Name or Material Albendazole sulfoxide
Grade HPLC