Learn More
Albendazole-sulfoxide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30315330
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315330 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30315330 Supplier Dr. Ehrenstorfer Supplier No. DREA10065400AL100
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Albendazole sulfoxide
CAS 54029-12-8
Analyte or Component Names Albendazole sulfoxide
Shipping Condition Room Temperature
Molecular Formula C12 H15 N3 O3 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Carbamic acid, [5-(propylsulfinyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), (±)-Albendazole sulfoxide, Albendazole oxide, Methyl [5-(propylsulfinyl)-1H-benzimidazol-2-yl]carbamate, RS 8852, Ricobendazole, Rycobendazole, Albendazole Imp. B (EP), SKF-77664, Ph Eur Albendazole Impurity B, Albendazole Impurity B
InChI Formula InChI=1S/C12H15N3O3S/c1-3-6-19(17)8-4-5-9-10(7-8)14-11(13-9)15-12(16)18-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)
SMILES CCCS(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1
IUPAC Name methyl N-(5-propylsulfinyl-1H-benzimidazol-2-yl)carbamate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 281.33
Quantity 1 mL
Formula Weight 281.0834 g/mol