missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKR7A2 Mouse anti-Human, Clone: 2E1E8, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | AKR7A2 |
|---|---|
| Clone | 2E1E8 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808785
|
Proteintech
66677-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898775
|
Proteintech
66677-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.Specifications
| AKR7A2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AFAR, AFAR1, AFB1 aldehyde reductase 1, AFB1 AR 1, AFB1 AR1, AKR7, AKR7A2, Aldoketoreductase 7, SSA reductase | |
| AKR7A2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2E1E8 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43488 | |
| 8574 | |
| AKR7A2 Fusion Protein Ag26105 | |
| Primary | |
| -20°C | |
| AKR7A2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title