missing translation for 'onlineSavingsMsg'
Learn More

AKR7A2 Mouse anti-Human, Clone: 2E1E8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66677-1-IG-20UL

 View more versions of this product

Product Code. 16808785

  • 145.00 € / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AKR7A2 is aldo-keto reductases, which are involved in the detoxification of aldehydes and ketones.
Specifications

Specifications

AKR7A2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43488
AKR7A2
AKR7A2 Fusion Protein Ag26105
20 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2E1E8
Unconjugated
AKR7A2
AFAR, AFAR1, AFB1 aldehyde reductase 1, AFB1 AR 1, AFB1 AR1, AKR7, AKR7A2, Aldoketoreductase 7, SSA reductase
Mouse
Protein G
RUO
8574
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.