Learn More
Memmert™ Additional Stainless Steel Grid for SN260plus, Reinforced Chamber Option
Product Code. 17588565 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN260plus Sterilizer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17588565 SN260plus Sterilizer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17588565 Supplier Memmert™ Supplier No. SN260plusE29766
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Additional grid features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
For Use With (Equipment) SN260plus Sterilizer
Product Type Reinforced Grid
Material Stainless Steel