Learn More
Memmert™ Additional Stainless Steel Grid for IN110, Reinforced Chamber Option
Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
Specifications
| Capacity (Metric) Shelf | 60 kg |
| For Use With (Equipment) | IN110 Incubator |
| Material | Stainless Steel |
| Type | Reinforced Grid |