Learn More
Memmert™ Additional Stainless Steel Grid, electro-polished, For Hcp105
Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
Includes
490 x 370 mm Glass Door
- Reinforced stainless steel grid
- For convection ovens
Specifications
Specifications
| For Use With (Equipment) | Models 110, 160 Incubator |
| Material | Stainless Steel |
| Type | Grid |