Learn More
Melford ACES
Click to view available options
Quantity:
100 g
500 g
5 kg
Unit Size:
100g
500g
5kg
Description
Zwitterionic buffer used in biochemistry and molecular biology research.
Zwitterionic buffer used in biochemistry and molecular biology research.
- pKa (25°C): 6.88
- The working pH range in aqueous solution is 6.1 - 7. 5
- Absorbance 280nm (5% water): <0.02
Specifications
Specifications
| Chemical Name or Material | ACES |
| CAS | 7365-82-4 |
| Quantity | 100 g |
| Molecular Formula | C4H10N2O4S |