missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Accucore™ C4 Reversed Phase HPLC Column, 2.6 μm, 300 μm x 5 mm
Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
5 mm
Unit Size:
Pack of 5
Description
- 150Å pore size solid core particles for fast biomoleucule separations
- Superb resolution at low backpressures
- Exceptionally rugged columns
- Significantly lower hydrophobic retention than C18
- Ideal for optimal retention of proteins and larger peptides
Specifications
Specifications
| Column Format | Trap Cartridge, 5/Pk. |
| Diameter (Metric) | 300 μm |
| Length (Metric) | 5 mm |
| Particle Size | 2.6 μm |
| pH | 2 to 9 |
| Pore Size | 150 Å |
| Temperature | 70°C |
| USP Type | L26 |
| Stationary Phase | C4 |
| Packing Material | Silica |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction