Learn More
Acclaim™ PepMap™ Fusica™ C18 Peptide Analysis HPLC Column, 5 μm, 0.3 mm x 250 mm
Product Code. 11322103 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
250 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11322103 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11322103 Supplier Thermo Scientific Chromatography Supplier No. 160300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Column Format: Capillary Column; Carbon Load: 15%; Pore Size: 100 Å; Column Phase: Reversed Phase
Offer high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ Acclaim 5μm PepMap 100 Fusica™ Capillary Columns.
  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Inside diameter: 300μm
TRUSTED_SUSTAINABILITY
Specifications
Column Format Capillary Column
Diameter (Metric) 0.3 mm
Flow Rate 4 μL/min
For Use With LC/MS, ESI-MS, MALDI-MS
Length (Metric) 250 mm
Max. Pressure 14500 psi (1000 bar)
Particle Shape Spherical
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m²/g
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 15%
Column Type Peptide Analysis
Phase Reversed Phase
Material Glass Lined Tubing