Learn More
Cytiva Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System
Product Code. 10488824 Shop All Cytiva Products
Click to view available options
Type:
Gel Supporting Screen
Masks
Unit Size:
Each
Pack of 5
Product Code. 10488824
Brand: Cytiva 18101043
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
  • Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
  • Efficient and reproducible transfers improve blotting results
  • Carry out all pretransfer steps in the unit
  • Precise vacuum control for optimal transfer conditions
  • RNA transfers in less than 4 hours
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Amersham VacuGene XL Vacuum Blotting System
Type Gel Supporting Screen