missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92667-0.02ml
This item is not returnable.
View return policy
Description
ABCA3 Polyclonal antibody specifically detects ABCA3 in Human, Rat samples. It is validated for Western Blot
Specifications
| ABCA3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ABC transporter 3, ABC3MGC72201, ABC-C, ABC-C transporter, ATP-binding cassette 3, ATP-binding cassette sub-family A member 3, ATP-binding cassette transporter 3, ATP-binding cassette, sub-family A (ABC1), member 3, EC 3.6.3, EC 3.6.3.17, EST111653, LBM180, MGC166979, SMDP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 43-203 of human ABCA3 (NP_001080.2). RLKIQSENVPNATIYPGQSIQELPLFFTFPPPGDTWELAYIPSHSDAAKTVTETVRRALVINMRVRGFPSEKDFEDYIRYDNCSSSVLAAVVFEHPFNHSKEPLPLAVKYHLRFSYTRRNYMWTQTGSFFLKETEGWHTTSLFPLFPNPGPREPTSPDGGE | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 21 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction