missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCA3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 487.00 €
Specifications
| Antigen | ABCA3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655212
|
Novus Biologicals
NBP2-92667-0.02ml |
0.02 mL |
216.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621562
|
Novus Biologicals
NBP2-92667-0.1ml |
0.1 mL |
487.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCA3 Polyclonal antibody specifically detects ABCA3 in Human, Rat samples. It is validated for Western BlotSpecifications
| ABCA3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 21 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ABC transporter 3, ABC3MGC72201, ABC-C, ABC-C transporter, ATP-binding cassette 3, ATP-binding cassette sub-family A member 3, ATP-binding cassette transporter 3, ATP-binding cassette, sub-family A (ABC1), member 3, EC 3.6.3, EC 3.6.3.17, EST111653, LBM180, MGC166979, SMDP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 43-203 of human ABCA3 (NP_001080.2). RLKIQSENVPNATIYPGQSIQELPLFFTFPPPGDTWELAYIPSHSDAAKTVTETVRRALVINMRVRGFPSEKDFEDYIRYDNCSSSVLAAVVFEHPFNHSKEPLPLAVKYHLRFSYTRRNYMWTQTGSFFLKETEGWHTTSLFPLFPNPGPREPTSPDGGE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title