Learn More
A-867744, MedChemExpress
Product Code. 16444998
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16444998
Brand: Medchem Express HY12149
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.
A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.
Specifications
Chemical Name or Material A-867744
CAS 1000279-69-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H19ClN2O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (248.21 mM)
SMILES O=S(C1=CC=C(C=C1)N2C(C)=C(C=C2C3=CC=C(C=C3)Cl)C(CC)=O)(N)=O
Molecular Weight (g/mol) 402.89
Formula Weight 402.89
Percent Purity 95.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation