Learn More
9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, Mikromol™
Description
9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one
Specifications
Specifications
| Chemical Name or Material | 9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one |
| CAS | 27387-31-1 |
| Analyte or Component Names | 9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H13 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ondansetron Hydrochloride Dihydrate Imp. C (EP), Ondansetron Imp. C (EP), Ondansetron USP Related Compound C, Ondansetron USP RC C, 9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one, GR 36948X, USP Ondanestron Related Compound C, Ph Eur Ondansetron Impurity C, methyl carbazolone (GR 36948X), Ondansetron Related Compound C, Ondansetron Hydrochloride Dihydrate Impurity C, Ondansetron Impurity C |
| InChI Formula | InChI=1S/C13H13NO/c1-14-10-6-3-2-5-9(10)13-11(14)7-4-8-12(13)15/h2-3,5-6H,4,7-8H2,1H3 |
| SMILES | Cn1c2CCCC(=O)c2c3ccccc13 |
| IUPAC Name | 9-methyl-2,3-dihydro-1H-carbazol-4-one |
| Show More |