Learn More
9-Chloro-11beta,17-dihydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl Methanesulfonate (Icometasone 21-Mesylate), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
9-Chloro-11beta,17-dihydroxy-16alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl Methanesulfonate (Icometasone 21-Mesylate)
Specifications
Specifications
| Chemical Name or Material | Icomethasone 21-Mesylate |
| CAS | 352315-75-4 |
| Analyte or Component Names | Icomethasone 21-Mesylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H31 Cl O7 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Pregna-1,4-diene-3,20-dione, 9-chloro-11,17-dihydroxy-16-methyl-21-[(methylsulfonyl)oxy]-, (11β,16α)-, (11β,16α)-9-Chloro-11,17-dihydroxy-16-methyl-21-[(methylsulfonyl)oxy]pregna-1,4-diene-3,20-dione, Icomethasone 21-mesylate, 9-Chloro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-21-yl Methanesulfonate, Mometasone Furoate Imp. N (EP), Mometasone Furoate Impurity N |
| InChI Formula | InChI=1S/C23H31ClO7S/c1-13-9-17-16-6-5-14-10-15(25)7-8-20(14,2)22(16,24)18(26)11-21(17,3)23(13,28)19(27)12-31-32(4,29)30/h7-8,10,13,16-18,26,28H,5-6,9,11-12H2,1-4H3/t13-,16+,17+,18+,20+,21+,22+,23+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COS(=O)(=O)C |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] methanesulfonate |
| Show More |