missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Chloro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-21-yl Furan-2-carboxylate (Icometasone 21-Furoate), Mikromol™
Description
9-Chloro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-21-yl Furan-2-carboxylate (Icometasone 21-Furoate)
Specifications
Specifications
| Chemical Name or Material | Icometasone 21-Furoate |
| Analyte or Component Names | Icometasone 21-Furoate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C27 H31 Cl O7 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 9-Chloro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregna-1,4-dien-21-yl Furan-2-carboxylate, Icometasone 21-Furoate, Mometasone Furoate Imp. P (EP) |
| InChI Formula | InChI=1S/C27H31ClO7/c1-15-11-19-18-7-6-16-12-17(29)8-9-24(16,2)26(18,28)21(30)13-25(19,3)27(15,33)22(31)14-35-23(32)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,30,33H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1 |
| SMILES | C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)c5occc5 |
| IUPAC Name | [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] furan-2-carboxylate |
| Molecular Weight (g/mol) | 502.9838 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?