Learn More
8-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one, Mikromol™
Description
8-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one
Specifications
Specifications
| Chemical Name or Material | 8-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]-benzimidazol-2(12H)-one |
| CAS | 125656-82-8 |
| Analyte or Component Names | 8-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]-benzimidazol-2(12H)-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H13 N3 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Omeprazole Imp. F (EP), Omeprazole USP Related Compound F and G Mixture, Omeprazole USP RC F and G Mixture, 8-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one, Omeprazole Related Compound F, Esomeprazole Sodium Impurity F, Omeprazole Impurity F, Esomeprazole Impurity F |
| InChI Formula | InChI=1S/C16H13N3O2S/c1-8-7-18-13(9(2)14(8)20)15(22)19-12-5-4-10(21-3)6-11(12)17-16(18)19/h4-7H,1-3H3 |
| SMILES | COc1ccc2c(c1)nc3N4C=C(C)C(=O)C(=C4C(=S)n23)C |
| IUPAC Name | 8-Methoxy-1,3-dimethyl-12-thioxopyrido[1',2':3,4]imidazo[1,2-a]benzimidazol-2(12H)-one |
| Show More |