missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline, 97%, Thermo Scientific™

Product Code. 11428727
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428727 5 g 5g
11418727 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428727 Supplier Thermo Scientific Alfa Aesar Supplier No. A19892.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18706-22-4
Molecular Formula C10H5ClF3NO
Molecular Weight (g/mol) 247.601
MDL Number MFCD00276589
InChI Key SBVZSZNYPRWWNN-UHFFFAOYSA-N
Synonym 8-chloro-4-hydroxy-2-trifluoromethyl quinoline, 8-chloro-2-trifluoromethyl quinolin-4-ol, 8-chloro-2-trifluoromethyl-1h-quinolin-4-one, 8-chloro-2-trifluoromethyl-4-quinolinol, 4 1h-quinolinone, 8-chloro-2-trifluoromethyl, maybridge1_004966, pubchem5940, acmc-1bvi2, 8-chloro-2-trifluoromethyl-quinolin-4-ol, 8-chloro-4-hydroxy-2-trifluoromethylquinoline
PubChem CID 2736611
IUPAC Name 8-chloro-2-(trifluoromethyl)-1H-quinolin-4-one
SMILES C1=CC2=C(C(=C1)Cl)NC(=CC2=O)C(F)(F)F

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
Formula Weight 247.61
Percent Purity 97%
Chemical Name or Material 8-Chloro-4-hydroxy-2-(trifluoromethyl)quinoline
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.